Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO11
Entrez Gene ID
80204
Gene Name
F-box protein 11
Gene Aliases
FBX11, PRMT9, UBR6, UG063H01, VIT1
Chromosome Location
Chr.2: 47806920 - 47905793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190274.1 NP_001177203.1 20 - 21 2512 96
NM_025133.4 NP_079409.3 20 - 21 2301 96
XM_005264572.4 XP_005264629.1 20 - 21 2506 96
XM_005264573.4 XP_005264630.1 20 - 21 2459 96
XM_017005015.1 XP_016860504.1 20 - 21 2459 96
XM_017005016.1 XP_016860505.1 20 - 21 2325 96
XM_017005017.1 XP_016860506.1 20 - 21 2325 96
GenBank mRNA AF264714.1 - 2 - 3 1447 96
AK025477.1 - 5 - 6 504 96
AK097734.1 - 9 - 10 1049 96
AK223592.1 - 20 - 21 2396 96
AK292877.1 - 20 - 21 2351 96
AK304140.1 - 20 - 21 2216 96
AK315108.1 - 20 - 21 2301 96
AL117620.1 - 3 - 4 283 96
AY827075.1 - 20 - 21 2439 96
BC012728.2 - 6 - 7 585 96
BC043258.1 - 20 - 21 2403 96
BC130445.1 - 20 - 21 2237 96
BC136480.1 - 20 - 21 2237 96

View Full Product Details