The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 7 - 8 | 1009 | 188 |
NM_025133.4 | NP_079409.3 | 7 - 8 | 798 | 188 | |
XM_005264572.4 | XP_005264629.1 | 7 - 8 | 1003 | 188 | |
XM_005264573.4 | XP_005264630.1 | 7 - 8 | 956 | 188 | |
XM_017005015.1 | XP_016860504.1 | 7 - 8 | 956 | 188 | |
XM_017005016.1 | XP_016860505.1 | 7 - 8 | 822 | 188 | |
XM_017005017.1 | XP_016860506.1 | 7 - 8 | 822 | 188 | |
GenBank mRNA | AF351618.1 | - | 7 - 8 | 811 | 188 |
AK022735.1 | - | 3 - 4 | 311 | 188 | |
AK223592.1 | - | 7 - 8 | 893 | 188 | |
AK292877.1 | - | 7 - 8 | 848 | 188 | |
AK297444.1 | - | 7 - 8 | 983 | 188 | |
AK304140.1 | - | 7 - 8 | 713 | 188 | |
AK315108.1 | - | 7 - 8 | 798 | 188 | |
AY827075.1 | - | 7 - 8 | 936 | 188 | |
BC043258.1 | - | 7 - 8 | 900 | 188 | |
BC130445.1 | - | 7 - 8 | 734 | 188 | |
BC136480.1 | - | 7 - 8 | 734 | 188 |