The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020869.3 | NP_065920.2 | 10 - 11 | 1548 | 107 |
GenBank mRNA | BX647348.1 | - | 10 - 11 | 1548 | 107 |