Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JRK
Entrez Gene ID
8629
Gene Name
Jrk helix-turn-helix protein
Gene Aliases
JH8, jerky
Chromosome Location
Chr.8: 142651501 - 142670132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001279352.1 NP_001266281.1 3 - 4 2229 81

View Full Product Details