The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114614.2 | NP_001108086.1 | 6 - 7 | 1014 | 104 |
GenBank mRNA | BC003610.2 | - | 6 - 7 | 907 | 104 |
BT006948.1 | - | 6 - 7 | 864 | 104 |