Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
Chromosome Location
Chr.8: 41653225 - 41896762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000037.3 NP_000028.3 10 - 11 1194 103
NM_001142446.1 NP_001135918.1 10 - 11 1491 103
NM_020475.2 NP_065208.2 10 - 11 1194 103
NM_020476.2 NP_065209.2 10 - 11 1194 103
NM_020477.2 NP_065210.2 10 - 11 1194 103
XM_005273476.4 XP_005273533.1 10 - 11 1468 103
XM_011544490.2 XP_011542792.1 11 - 12 1390 103
XM_011544491.2 XP_011542793.1 11 - 12 1389 103
XM_011544494.2 XP_011542796.1 11 - 12 1389 103
XM_011544495.2 XP_011542797.1 11 - 12 1388 103
XM_011544496.2 XP_011542798.1 11 - 12 1390 103
XM_011544500.2 XP_011542802.1 10 - 11 1296 103
XM_011544501.2 XP_011542803.1 10 - 11 1296 103
XM_011544502.2 XP_011542804.1 10 - 11 1296 103
XM_011544503.2 XP_011542805.1 11 - 12 1387 103
XM_011544504.2 XP_011542806.1 10 - 11 1296 103
XM_011544505.2 XP_011542807.1 10 - 11 1296 103
XM_017013319.1 XP_016868808.1 11 - 12 1389 103
XM_017013320.1 XP_016868809.1 11 - 12 1391 103
XM_017013321.1 XP_016868810.1 11 - 12 1334 103
XM_017013322.1 XP_016868811.1 10 - 11 1296 103
XM_017013323.1 XP_016868812.1 11 - 12 1310 103
XM_017013324.1 XP_016868813.1 10 - 11 1468 103
XM_017013325.1 XP_016868814.1 10 - 11 1235 103
XM_017013326.1 XP_016868815.1 10 - 11 1228 103
XM_017013327.1 XP_016868816.1 11 - 12 1389 103
XM_017013328.1 XP_016868817.1 11 - 12 1389 103
XM_017013329.1 XP_016868818.1 10 - 11 1466 103
GenBank mRNA AB209418.1 - 10 - 11 1491 103
M28880.1 - 10 - 11 1194 103
X16609.1 - 10 - 11 1194 103

View Full Product Details