Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANK1
- Entrez Gene ID
-
286
- Gene Name
- ankyrin 1
- Gene Aliases
-
ANK, SPH1, SPH2
- Chromosome Location
-
Chr.8: 41653225 - 41896762 on Build GRCh38
View Full Product Details