Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
Chromosome Location
Chr.8: 41653225 - 41896762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000037.3 NP_000028.3 6 - 7 697 81
NM_001142446.1 NP_001135918.1 6 - 7 994 81
NM_020475.2 NP_065208.2 6 - 7 697 81
NM_020476.2 NP_065209.2 6 - 7 697 81
NM_020477.2 NP_065210.2 6 - 7 697 81
XM_005273476.4 XP_005273533.1 6 - 7 971 81
XM_011544490.2 XP_011542792.1 7 - 8 893 81
XM_011544491.2 XP_011542793.1 7 - 8 892 81
XM_011544494.2 XP_011542796.1 7 - 8 892 81
XM_011544495.2 XP_011542797.1 7 - 8 891 81
XM_011544496.2 XP_011542798.1 7 - 8 893 81
XM_011544500.2 XP_011542802.1 6 - 7 799 81
XM_011544501.2 XP_011542803.1 6 - 7 799 81
XM_011544502.2 XP_011542804.1 6 - 7 799 81
XM_011544503.2 XP_011542805.1 7 - 8 890 81
XM_011544504.2 XP_011542806.1 6 - 7 799 81
XM_011544505.2 XP_011542807.1 6 - 7 799 81
XM_017013319.1 XP_016868808.1 7 - 8 892 81
XM_017013320.1 XP_016868809.1 7 - 8 894 81
XM_017013321.1 XP_016868810.1 7 - 8 837 81
XM_017013322.1 XP_016868811.1 6 - 7 799 81
XM_017013323.1 XP_016868812.1 7 - 8 813 81
XM_017013324.1 XP_016868813.1 6 - 7 971 81
XM_017013325.1 XP_016868814.1 6 - 7 738 81
XM_017013326.1 XP_016868815.1 6 - 7 731 81
XM_017013327.1 XP_016868816.1 7 - 8 892 81
XM_017013328.1 XP_016868817.1 7 - 8 892 81
XM_017013329.1 XP_016868818.1 6 - 7 969 81
GenBank mRNA AB209418.1 - 6 - 7 994 81
M28880.1 - 6 - 7 697 81
X16609.1 - 6 - 7 697 81

View Full Product Details