Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS10
Entrez Gene ID
55174
Gene Name
integrator complex subunit 10
Gene Aliases
C8orf35, INT10
Chromosome Location
Chr.8: 19817149 - 19852078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018142.2 NP_060612.2 8 - 9 1139 153
XM_005273555.3 XP_005273612.1 8 - 9 1397 153
XM_005273556.3 XP_005273613.1 8 - 9 1181 153
XM_005273557.3 XP_005273614.1 8 - 9 1181 153
XM_006716355.2 XP_006716418.1 8 - 9 1286 153
XM_017013606.1 XP_016869095.1 8 - 9 1181 153
XM_017013607.1 XP_016869096.1 8 - 9 1397 153
GenBank mRNA AK001431.1 - 8 - 9 1059 153
BC001915.1 - 6 - 7 776 153
BC006209.2 - 6 - 7 776 153
BC051282.1 - 6 - 7 790 153
CR457262.1 - 5 - 6 669 153

View Full Product Details