The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166237.1 | NP_001159709.1 | 8 - 9 | 1386 | 70 |
NM_024736.6 | NP_079012.3 | 5 - 6 | 832 | 70 | |
XM_011517301.1 | XP_011515603.1 | 4 - 5 | 1434 | 70 | |
GenBank mRNA | AF318341.1 | - | 6 - 7 | 870 | 70 |
AK022212.1 | - | 5 - 6 | 791 | 70 | |
AK093858.1 | - | 2 - 3 | 836 | 70 | |
AK096216.1 | - | 5 - 6 | 579 | 70 | |
AK291817.1 | - | 8 - 9 | 1386 | 70 | |
AY008304.1 | - | 5 - 6 | 821 | 70 | |
BC008904.2 | - | 5 - 6 | 817 | 70 | |
BC069000.1 | - | 5 - 6 | 772 | 70 |