The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166237.1 | NP_001159709.1 | 11 - 12 | 1701 | 115 |
NM_024736.6 | NP_079012.3 | 8 - 9 | 1147 | 115 | |
XM_011517301.1 | XP_011515603.1 | 7 - 8 | 1749 | 115 | |
GenBank mRNA | AF318341.1 | - | 9 - 10 | 115 | |
AK022212.1 | - | 8 - 9 | 1106 | 115 | |
AK096216.1 | - | 8 - 9 | 894 | 115 | |
AK123985.1 | - | 6 - 7 | 1595 | 115 | |
AK291817.1 | - | 11 - 12 | 1701 | 115 | |
BC008904.2 | - | 8 - 9 | 1132 | 115 | |
BC069000.1 | - | 8 - 9 | 1087 | 115 |