The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284220.1 | NP_001271149.1 | 13 - 14 | 1611 | 85 |
NM_001284222.1 | NP_001271151.1 | 14 - 15 | 1646 | 85 | |
NM_001284224.1 | NP_001271153.1 | 12 - 13 | 1508 | 85 | |
NM_001284225.1 | NP_001271154.1 | 12 - 13 | 1547 | 85 | |
NM_001284226.1 | NP_001271155.1 | 13 - 14 | 1572 | 85 | |
NM_018091.5 | NP_060561.3 | 14 - 15 | 1711 | 85 | |
XM_006716354.2 | XP_006716417.1 | 13 - 14 | 1601 | 85 | |
GenBank mRNA | AK001284.1 | - | 14 - 15 | 1494 | 85 |
AK022626.1 | - | 14 - 15 | 1667 | 85 | |
AK223047.1 | - | 13 - 14 | 1497 | 85 | |
AK293424.1 | - | 12 - 13 | 1474 | 85 | |
AK295574.1 | - | 13 - 14 | 1500 | 85 | |
AK296475.1 | - | 13 - 14 | 1566 | 85 | |
AK298265.1 | - | 13 - 14 | 1462 | 85 | |
AK302140.1 | - | 12 - 13 | 1473 | 85 | |
AK315985.1 | - | 12 - 13 | 1451 | 85 | |
AK316053.1 | - | 13 - 14 | 1515 | 85 | |
AL834273.1 | - | 14 - 15 | 1569 | 85 | |
BC001240.1 | - | 14 - 15 | 1601 | 85 | |
BX648011.1 | - | 14 - 15 | 1645 | 85 |