The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001255978.1 | NP_001242907.1 | 2 - 3 | 508 | 105 |
NM_001255979.1 | NP_001242908.1 | 2 - 3 | 294 | 105 | |
NM_130898.3 | NP_570968.1 | 2 - 3 | 528 | 105 | |
GenBank mRNA | AB052778.1 | - | 2 - 3 | 446 | 105 |
AF394167.1 | - | 2 - 3 | 256 | 105 | |
AF468007.1 | - | 2 - 3 | 295 | 105 | |
AK304259.1 | - | 2 - 3 | 240 | 105 | |
AY049977.1 | - | 2 - 3 | 444 | 105 | |
BC038962.1 | - | 2 - 3 | 430 | 105 |