The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004980.4 | NP_004971.2 | 2 - 3 | 1591 | 80 |
NM_172198.2 | NP_751948.1 | 2 - 3 | 1591 | 80 | |
XM_005270851.4 | XP_005270908.1 | 2 - 3 | 1256 | 80 | |
XM_006710629.3 | XP_006710692.1 | 2 - 3 | 1566 | 80 | |
XM_006710630.3 | XP_006710693.1 | 2 - 3 | 1256 | 80 | |
XM_006710631.3 | XP_006710694.1 | 2 - 3 | 1256 | 80 | |
XM_017001244.1 | XP_016856733.1 | 2 - 3 | 1581 | 80 | |
GenBank mRNA | AF205856.1 | - | 1 - 2 | 1111 | 80 |
AF205857.1 | - | 1 - 2 | 1111 | 80 | |
BC113475.1 | - | 1 - 2 | 1133 | 80 | |
BC113477.1 | - | 1 - 2 | 80 | ||
BC143390.1 | - | 1 - 2 | 1133 | 80 |