The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261464.1 | NP_001248393.1 | 1 - 2 | 196 | 87 |
GenBank mRNA | AK296925.1 | - | 1 - 2 | 117 | 87 |