The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014712.2 | NP_055527.1 | 18 - 19 | 5637 | 67 |
XM_005255723.1 | XP_005255780.1 | 18 - 19 | 5121 | 67 | |
XM_006721106.3 | XP_006721169.1 | 18 - 19 | 5042 | 67 | |
XM_017023909.1 | XP_016879398.1 | 18 - 19 | 6274 | 67 | |
GenBank mRNA | AB002337.2 | - | 18 - 19 | 5637 | 67 |
BC027450.1 | - | 5 - 6 | 1237 | 67 |