Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPH
Entrez Gene ID
5630
Gene Name
peripherin
Gene Aliases
NEF4, PRPH1
Chromosome Location
Chr.12: 49295013 - 49298698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006262.3 NP_006253.2 1 - 2 622 147
XM_005269025.1 XP_005269082.1 1 - 2 735 147
GenBank mRNA AK125587.1 - 1 - 2 2046 147
BC032703.1 - 1 - 2 582 147

View Full Product Details