Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
51093
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, CGI-41, METTL25B
Chromosome Location
Chr.1: 156728471 - 156736960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142560.1 NP_001136032.1 2 - 3 868 109
NM_015997.3 NP_057081.3 2 - 3 868 109
XM_005245234.4 XP_005245291.1 2 - 3 653 109
XM_005245236.4 XP_005245293.1 2 - 3 653 109
XM_005245237.4 XP_005245294.1 2 - 3 653 109
XM_011509600.1 XP_011507902.1 2 - 3 205 109
XM_017001404.1 XP_016856893.1 2 - 3 788 109
GenBank mRNA AF151800.1 - 2 - 3 458 109
AK000412.1 - 3 - 4 684 109
BC011382.1 - 2 - 3 853 109

View Full Product Details