The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142560.1 | NP_001136032.1 | 3 - 4 | 1068 | 71 |
NM_015997.3 | NP_057081.3 | 3 - 4 | 1068 | 71 | |
XM_005245234.4 | XP_005245291.1 | 3 - 4 | 853 | 71 | |
XM_005245235.4 | XP_005245292.1 | 2 - 3 | 712 | 71 | |
XM_005245236.4 | XP_005245293.1 | 3 - 4 | 853 | 71 | |
XM_005245237.4 | XP_005245294.1 | 3 - 4 | 853 | 71 | |
XM_011509600.1 | XP_011507902.1 | 3 - 4 | 405 | 71 | |
XM_017001404.1 | XP_016856893.1 | 3 - 4 | 988 | 71 | |
XM_017001405.1 | XP_016856894.1 | 3 - 4 | 933 | 71 | |
XM_017001406.1 | XP_016856895.1 | 3 - 4 | 939 | 71 | |
GenBank mRNA | AK000412.1 | - | 4 - 5 | 884 | 71 |
BC011382.1 | - | 3 - 4 | 1053 | 71 |