The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171818.1 | NP_001165289.1 | 1 - 2 | 124 | 94 |
NM_001171819.1 | NP_001165290.1 | 1 - 2 | 124 | 94 | |
NM_001171820.1 | NP_001165291.1 | 1 - 2 | 124 | 94 | |
NM_006238.4 | NP_006229.1 | 1 - 2 | 124 | 94 | |
NM_177435.2 | NP_803184.1 | 1 - 2 | 124 | 94 | |
XM_006715120.1 | XP_006715183.1 | 1 - 2 | 111 | 94 | |
XM_011514707.1 | XP_011513009.1 | 1 - 2 | 111 | 94 | |
XM_011514709.1 | XP_011513011.1 | 1 - 2 | 111 | 94 | |
XM_011514710.1 | XP_011513012.1 | 1 - 2 | 111 | 94 | |
XM_017010971.1 | XP_016866460.1 | 1 - 2 | 111 | 94 | |
XM_017010972.1 | XP_016866461.1 | 1 - 2 | 111 | 94 | |
XM_017010973.1 | XP_016866462.1 | 1 - 2 | 111 | 94 | |
GenBank mRNA | AB099507.1 | - | 1 - 2 | 87 | 95 |
AK122614.1 | - | 1 - 2 | 117 | 94 | |
AK291661.1 | - | 1 - 2 | 83 | 94 | |
AK296425.1 | - | 1 - 2 | 83 | 94 | |
AK304878.1 | - | 1 - 2 | 87 | 94 | |
BC002715.2 | - | 1 - 2 | 73 | 94 | |
BC007578.1 | - | 1 - 2 | 73 | 94 |