Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PPARD
- Entrez Gene ID
-
5467
- Gene Name
- peroxisome proliferator activated receptor delta
- Gene Aliases
-
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
- Chromosome Location
-
Chr.6: 35342558 - 35428191 on Build GRCh38
View Full Product Details