Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
Chromosome Location
Chr.22: 20080232 - 20111877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190326.1 NP_001177255.1 2 - 3 1148 69
NM_022720.6 NP_073557.3 2 - 3 1148 69
GenBank mRNA AK025539.1 - 1 - 2 112 69
AK222796.1 - 1 - 2 112 69
AK313357.1 - 2 - 3 1099 69
BC009984.2 - 2 - 3 1069 69
BC037564.1 - 2 - 3 1098 69
BC078147.1 - 1 - 2 862 69
CR456356.1 - 1 - 2 806 69

View Full Product Details