The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282129.1 | NP_001269058.1 | 3 - 4 | 416 | 66 |
NM_001282130.1 | NP_001269059.1 | 2 - 3 | 335 | 66 | |
NM_033389.3 | NP_203747.2 | 2 - 3 | 147 | 66 | |
XM_005258058.3 | XP_005258115.1 | 4 - 5 | 309 | 66 | |
XM_011525402.2 | XP_011523704.1 | 3 - 4 | 144 | 66 | |
XM_011525407.1 | XP_011523709.1 | 1 - 2 | 44 | 66 | |
XM_017025251.1 | XP_016880740.1 | 3 - 4 | 355 | 66 | |
XM_017025252.1 | XP_016880741.1 | 2 - 3 | 335 | 66 | |
XM_017025253.1 | XP_016880742.1 | 3 - 4 | 355 | 66 | |
XM_017025254.1 | XP_016880743.1 | 3 - 4 | 355 | 66 | |
XM_017025255.1 | XP_016880744.1 | 3 - 4 | 355 | 66 | |
GenBank mRNA | AB072358.1 | - | 2 - 3 | 107 | 66 |
AB072359.1 | - | 2 - 3 | 318 | 66 | |
AB099290.1 | - | 2 - 3 | 107 | 66 | |
AK296413.1 | - | 3 - 4 | 281 | 66 | |
AK308887.1 | - | 2 - 3 | 214 | 66 | |
BC008941.2 | - | 2 - 3 | 259 | 66 | |
BC068223.1 | - | 2 - 3 | 259 | 66 | |
DA749059.1 | - | 2 - 3 | 336 | 66 |