Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
Chromosome Location
Chr.17: 29625938 - 29930239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282130.1 NP_001269059.1 7 - 8 761 137
NM_001282131.1 NP_001269060.1 6 - 7 709 137
XM_017025255.1 XP_016880744.1 8 - 9 781 137
GenBank mRNA AB072359.1 - 7 - 8 137
BC008941.2 - 7 - 8 685 137
BC011636.2 - 6 - 7 667 137
BC068223.1 - 7 - 8 685 137

View Full Product Details