Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HABP4
- Entrez Gene ID
-
22927
- Gene Name
- hyaluronan binding protein 4
- Gene Aliases
-
IHABP-4, IHABP4, Ki-1/57, SERBP1L
- Chromosome Location
-
Chr.9: 96450155 - 96491336 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC018788.1 |
- |
2 - 3 |
597 |
127 |
View Full Product Details