The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267045.1 | NP_001253974.1 | 7 - 8 | 1108 | 62 |
NM_005789.3 | NP_005780.2 | 5 - 6 | 781 | 62 | |
NM_176863.2 | NP_789839.1 | 5 - 6 | 781 | 62 | |
XM_011524177.2 | XP_011522479.1 | 5 - 6 | 790 | 62 | |
GenBank mRNA | AK057221.1 | - | 7 - 8 | 947 | 62 |
AK074999.1 | - | 5 - 6 | 478 | 62 | |
AK307110.1 | - | 4 - 5 | 1182 | 62 | |
AK314347.1 | - | 5 - 6 | 524 | 62 | |
BC001423.2 | - | 5 - 6 | 508 | 62 | |
BC002684.1 | - | 5 - 6 | 487 | 62 | |
BC008020.1 | - | 5 - 6 | 439 | 62 | |
BG708969.1 | - | 5 - 6 | 522 | 62 | |
BT019386.1 | - | 5 - 6 | 291 | 62 | |
BX640801.1 | - | 7 - 8 | 835 | 62 | |
CR541745.1 | - | 5 - 6 | 291 | 62 | |
EU794653.1 | - | 5 - 6 | 508 | 62 | |
FJ200488.1 | - | 5 - 6 | 291 | 62 | |
U11292.1 | - | 5 - 6 | 453 | 62 |