The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206729.1 | NP_001193658.1 | 7 - 8 | 782 | 107 |
NM_005465.4 | NP_005456.1 | 6 - 7 | 741 | 107 | |
NM_181690.2 | NP_859029.1 | 6 - 7 | 741 | 107 | |
XM_005272994.4 | XP_005273051.1 | 7 - 8 | 966 | 107 | |
XM_005272995.2 | XP_005273052.1 | 7 - 8 | 772 | 107 | |
XM_006711726.3 | XP_006711789.1 | 7 - 8 | 966 | 107 | |
XM_011544012.2 | XP_011542314.1 | 7 - 8 | 966 | 107 | |
XM_011544013.2 | XP_011542315.1 | 7 - 8 | 966 | 107 | |
XM_016999985.1 | XP_016855474.1 | 8 - 9 | 914 | 107 | |
GenBank mRNA | AF085234.1 | - | 6 - 7 | 676 | 107 |
AF124141.1 | - | 6 - 7 | 641 | 107 | |
AF135794.1 | - | 6 - 7 | 629 | 107 | |
AJ245709.1 | - | 6 - 7 | 639 | 107 | |
AK308052.1 | - | 6 - 7 | 667 | 107 | |
AL117525.1 | - | 7 - 8 | 782 | 107 | |
AY005799.1 | - | 6 - 7 | 702 | 107 | |
BC020479.1 | - | 6 - 7 | 675 | 107 | |
BC121154.2 | - | 6 - 7 | 712 | 107 |