Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF11
Entrez Gene ID
9169
Gene Name
SR-related CTD associated factor 11
Gene Aliases
CASP11, SFRS2IP, SIP1, SRRP129, SRSF2IP
Chromosome Location
Chr.12: 45919131 - 45992040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004719.2 NP_004710.2 9 - 10 1018 106
XM_005269230.2 XP_005269287.2 9 - 10 908 106
XM_011538984.2 XP_011537286.1 10 - 11 2537 106
XM_011538985.1 XP_011537287.1 9 - 10 852 106
XM_017020217.1 XP_016875706.1 10 - 11 896 106
XM_017020218.1 XP_016875707.1 10 - 11 896 106
XM_017020219.1 XP_016875708.1 10 - 11 1095 106
XM_017020220.1 XP_016875709.1 10 - 11 896 106
XM_017020221.1 XP_016875710.1 8 - 9 1225 106
XM_017020222.1 XP_016875711.1 7 - 8 1195 106
XM_017020223.1 XP_016875712.1 10 - 11 1118 106
GenBank mRNA AF030234.1 - 9 - 10 966 106
AK026473.1 - 10 - 11 1018 106
AK302999.1 - 9 - 10 943 106
AK314093.1 - 3 - 4 281 106
BC017465.2 - 6 - 7 467 106
BC033589.1 - 5 - 6 423 106
BC040951.2 - 1 - 2 99 106

View Full Product Details