Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
Chromosome Location
Chr.8: 130052104 - 130443848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247996.1 NP_001234925.1 27 - 28 2978 123
NM_018482.3 NP_060952.2 26 - 27 2876 123
XM_005250925.1 XP_005250982.1 27 - 28 2885 123
XM_006716563.3 XP_006716626.1 27 - 28 2704 123
XM_006716564.1 XP_006716627.1 28 - 29 2863 123
XM_006716565.3 XP_006716628.1 26 - 27 2700 123
XM_011517052.2 XP_011515354.1 27 - 28 2701 123
XM_011517053.1 XP_011515355.1 27 - 28 2907 123
XM_017013467.1 XP_016868956.1 25 - 26 2722 123
XM_017013468.1 XP_016868957.1 26 - 27 2946 123
GenBank mRNA AB033075.1 - 20 - 21 2109 123
BC137135.1 - 25 - 26 2673 123
BC144493.1 - 26 - 27 2775 123
BX537768.1 - 19 - 20 2129 123

View Full Product Details