Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM71
Entrez Gene ID
137835
Gene Name
transmembrane protein 71
Gene Aliases
-
Chromosome Location
Chr.8: 132709945 - 132760668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144649.2 NP_653250.2 4 - 5 542 78
GenBank mRNA AK057631.1 - 4 - 5 456 78
BC062592.1 - 4 - 5 437 78

View Full Product Details