Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNPEPL1
Entrez Gene ID
57140
Gene Name
arginyl aminopeptidase like 1
Gene Aliases
-
Chromosome Location
Chr.2: 240568526 - 240578732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018226.4 NP_060696.4 4 - 5 932 62
XM_005247036.4 XP_005247093.1 4 - 5 993 62
GenBank mRNA AF300795.1 - 4 - 5 638 62
BC017301.2 - 1 - 2 84 62
BC067258.1 - 4 - 5 507 62
BC082975.1 - 4 - 5 832 62

View Full Product Details