Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 24 - 25 2516 150
NM_001320705.1 NP_001307634.1 24 - 25 2516 150
NM_004321.6 NP_004312.2 22 - 23 2302 150
XM_005247022.1 XP_005247079.1 24 - 25 2427 150
XM_005247024.1 XP_005247081.1 23 - 24 2400 150
XM_005247027.1 XP_005247084.1 23 - 24 2400 150
XM_005247028.2 XP_005247085.1 23 - 24 2403 150
XM_006712605.1 XP_006712668.1 23 - 24 2400 150
XM_011511364.1 XP_011509666.1 24 - 25 2439 150
XM_011511365.1 XP_011509667.1 24 - 25 2427 150
XM_011511366.2 XP_011509668.1 14 - 15 1499 150
XM_011511367.1 XP_011509669.1 14 - 15 1429 150
XM_017004383.1 XP_016859872.1 24 - 25 2502 150
XM_017004384.1 XP_016859873.1 24 - 25 2514 150
XM_017004385.1 XP_016859874.1 24 - 25 2502 150
XM_017004386.1 XP_016859875.1 24 - 25 2502 150
XM_017004387.1 XP_016859876.1 24 - 25 2502 150
XM_017004388.1 XP_016859877.1 24 - 25 2502 150
XM_017004389.1 XP_016859878.1 24 - 25 2502 150
XM_017004390.1 XP_016859879.1 23 - 24 2400 150
XM_017004391.1 XP_016859880.1 23 - 24 2281 150
GenBank mRNA AB210029.1 - 24 - 25 2516 150
AB290172.1 - 24 - 25 2516 150
BC111780.1 - 22 - 23 2270 150
BC111799.1 - 22 - 23 2270 150
BF946744.1 - 2 - 3 150
KJ534875.1 - 22 - 23 2242 150

View Full Product Details