Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 11 - 12 1207 73
NM_001320705.1 NP_001307634.1 11 - 12 1207 73
NM_004321.6 NP_004312.2 10 - 11 1020 73
XM_005247022.1 XP_005247079.1 11 - 12 1118 73
XM_005247024.1 XP_005247081.1 11 - 12 1118 73
XM_005247027.1 XP_005247084.1 11 - 12 1118 73
XM_005247028.2 XP_005247085.1 11 - 12 1121 73
XM_006712605.1 XP_006712668.1 11 - 12 1118 73
XM_011511364.1 XP_011509666.1 11 - 12 1130 73
XM_011511365.1 XP_011509667.1 11 - 12 1118 73
XM_011511366.2 XP_011509668.1 2 - 3 217 73
XM_011511367.1 XP_011509669.1 2 - 3 147 73
XM_017004383.1 XP_016859872.1 11 - 12 1118 73
XM_017004384.1 XP_016859873.1 11 - 12 1130 73
XM_017004385.1 XP_016859874.1 11 - 12 1118 73
XM_017004386.1 XP_016859875.1 11 - 12 1118 73
XM_017004387.1 XP_016859876.1 11 - 12 1118 73
XM_017004388.1 XP_016859877.1 11 - 12 1118 73
XM_017004389.1 XP_016859878.1 11 - 12 1118 73
XM_017004390.1 XP_016859879.1 11 - 12 1118 73
XM_017004391.1 XP_016859880.1 10 - 11 972 73
GenBank mRNA AB210029.1 - 11 - 12 1207 73
AB290172.1 - 11 - 12 1207 73
AL706949.1 - 5 - 6 331 73
BC111780.1 - 10 - 11 988 73
BC111799.1 - 10 - 11 988 73
KJ534875.1 - 10 - 11 960 73

View Full Product Details