Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location
Chr.2: 240713764 - 240821025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244008.1 NP_001230937.1 19 - 20 1933 100
NM_001320705.1 NP_001307634.1 19 - 20 1933 100
NM_004321.6 NP_004312.2 17 - 18 1719 100
XM_005247022.1 XP_005247079.1 19 - 20 1844 100
XM_005247024.1 XP_005247081.1 18 - 19 1817 100
XM_005247027.1 XP_005247084.1 18 - 19 1817 100
XM_005247028.2 XP_005247085.1 18 - 19 1820 100
XM_006712605.1 XP_006712668.1 18 - 19 1817 100
XM_011511364.1 XP_011509666.1 19 - 20 1856 100
XM_011511365.1 XP_011509667.1 19 - 20 1844 100
XM_011511366.2 XP_011509668.1 9 - 10 916 100
XM_011511367.1 XP_011509669.1 9 - 10 846 100
XM_017004383.1 XP_016859872.1 19 - 20 1919 100
XM_017004384.1 XP_016859873.1 19 - 20 1931 100
XM_017004385.1 XP_016859874.1 19 - 20 1919 100
XM_017004386.1 XP_016859875.1 19 - 20 1919 100
XM_017004387.1 XP_016859876.1 19 - 20 1919 100
XM_017004388.1 XP_016859877.1 19 - 20 1919 100
XM_017004389.1 XP_016859878.1 19 - 20 1919 100
XM_017004390.1 XP_016859879.1 18 - 19 1817 100
XM_017004391.1 XP_016859880.1 18 - 19 1698 100
GenBank mRNA AB210029.1 - 19 - 20 1933 100
AB290172.1 - 19 - 20 1933 100
BC111780.1 - 17 - 18 1687 100
BC111799.1 - 17 - 18 1687 100
BF879579.1 - 1 - 2 109 99
KJ534875.1 - 17 - 18 1659 100

View Full Product Details