Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KIF1A
- Entrez Gene ID
-
547
- Gene Name
- kinesin family member 1A
- Gene Aliases
-
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
- Chromosome Location
-
Chr.2: 240713764 - 240821025 on Build GRCh38
View Full Product Details