The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203394.2 | NP_976328.2 | 7 - 8 | 1362 | 122 |
XM_011537969.2 | XP_011536271.1 | 6 - 7 | 925 | 122 | |
GenBank mRNA | AK097677.1 | - | 5 - 6 | 953 | 122 |
BC136366.1 | - | 7 - 8 | 1201 | 122 | |
BC136367.1 | - | 7 - 8 | 1201 | 122 |