Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP54
Entrez Gene ID
144535
Gene Name
cilia and flagella associated protein 54
Gene Aliases
C12orf55, C12orf63
Chromosome Location
Chr.12: 96489566 - 96875555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306084.1 NP_001293013.1 21 - 22 2931 134
XM_011539069.2 XP_011537371.1 21 - 22 2936 134
XM_011539070.2 XP_011537372.1 21 - 22 2936 134
XM_011539071.2 XP_011537373.1 21 - 22 2936 134
XM_011539072.2 XP_011537374.1 21 - 22 2936 134
XM_011539073.2 XP_011537375.1 21 - 22 2936 134
XM_011539074.2 XP_011537376.1 21 - 22 2936 134
XM_011539076.2 XP_011537378.1 21 - 22 2936 134
XM_011539077.2 XP_011537379.1 20 - 21 3086 134
XM_011539078.2 XP_011537380.1 21 - 22 2936 134
XM_011539079.1 XP_011537381.1 17 - 18 2182 134
XM_011539081.2 XP_011537383.1 21 - 22 2936 134
XM_011539083.2 XP_011537385.1 21 - 22 2936 134
XM_011539084.2 XP_011537386.1 21 - 22 2936 134
XM_011539085.1 XP_011537387.1 6 - 7 757 134
XM_017018866.1 XP_016874355.1 21 - 22 2936 134
XM_017018867.1 XP_016874356.1 10 - 11 1188 134
GenBank mRNA AK091789.1 - 7 - 8 866 134

View Full Product Details