The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303511.1 | NP_001290440.1 | 2 - 3 | 209 | 77 |
NM_005230.3 | NP_005221.2 | 4 - 5 | 1446 | 77 | |
XM_006719275.3 | XP_006719338.1 | 5 - 6 | 1543 | 77 | |
GenBank mRNA | AK312694.1 | - | 4 - 5 | 1357 | 77 |
AL705041.1 | - | 2 - 3 | 209 | 77 | |
BC017371.1 | - | 4 - 5 | 1328 | 77 | |
CR542251.1 | - | 3 - 4 | 1120 | 77 | |
Z36715.1 | - | 4 - 5 | 1399 | 77 |