The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014255.5 | NP_055070.1 | 5 - 6 | 1053 | 75 |
GenBank mRNA | AB015631.1 | - | 5 - 6 | 648 | 75 |
AK312921.1 | - | 5 - 6 | 750 | 75 | |
AY032624.1 | - | 5 - 6 | 922 | 75 | |
AY359102.1 | - | 5 - 6 | 1046 | 75 | |
BC065015.1 | - | 5 - 6 | 688 | 75 |