The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023068.3 | NP_075556.1 | 16 - 17 | 4336 | 107 |
XM_006723610.3 | XP_006723673.1 | 17 - 18 | 4531 | 107 | |
XM_011529324.2 | XP_011527626.1 | 17 - 18 | 4528 | 107 | |
XM_011529325.2 | XP_011527627.1 | 17 - 18 | 4534 | 107 | |
XM_011529326.2 | XP_011527628.1 | 17 - 18 | 4530 | 107 | |
XM_011529327.2 | XP_011527629.1 | 16 - 17 | 4464 | 107 | |
XM_011529328.2 | XP_011527630.1 | 17 - 18 | 4476 | 107 | |
XM_011529329.1 | XP_011527631.1 | 16 - 17 | 5717 | 107 | |
XM_011529330.1 | XP_011527632.1 | 15 - 16 | 5459 | 107 | |
XM_011529331.1 | XP_011527633.1 | 16 - 17 | 5635 | 107 | |
XM_011529332.1 | XP_011527634.1 | 15 - 16 | 3921 | 107 | |
GenBank mRNA | AF230073.1 | - | 16 - 17 | 4336 | 107 |
AK024459.1 | - | 3 - 4 | 776 | 107 | |
AK024462.1 | - | 8 - 9 | 2140 | 107 | |
AK024479.1 | - | 3 - 4 | 776 | 107 | |
AK122586.1 | - | 5 - 6 | 4307 | 107 | |
AL833688.1 | - | 1 - 2 | 71 | 107 | |
BC141884.1 | - | 16 - 17 | 4336 | 107 |