The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_023068.3 | NP_075556.1 | 4 - 5 | 976 | 106 | |
XM_006723610.3 | XP_006723673.1 | 5 - 6 | 1171 | 106 | ||
XM_011529324.2 | XP_011527626.1 | 5 - 6 | 1168 | 106 | ||
XM_011529325.2 | XP_011527627.1 | 5 - 6 | 1174 | 106 | ||
XM_011529326.2 | XP_011527628.1 | 5 - 6 | 1170 | 106 | ||
XM_011529327.2 | XP_011527629.1 | 4 - 5 | 1104 | 106 | ||
XM_011529328.2 | XP_011527630.1 | 5 - 6 | 1116 | 106 | ||
XM_011529329.1 | XP_011527631.1 | 4 - 5 | 2357 | 106 | ||
XM_011529330.1 | XP_011527632.1 | 4 - 5 | 2357 | 106 | ||
XM_011529331.1 | XP_011527633.1 | 4 - 5 | 2275 | 106 | ||
XM_011529332.1 | XP_011527634.1 | 3 - 4 | 561 | 106 | ||
XM_011529333.2 | XP_011527635.1 | 4 - 5 | 2357 | 106 | ||
GenBank mRNA | AF230073.1 | - | 4 - 5 | 976 | 106 | |
AY927519.1 | - | LIT3529 AY927519 |
3 - 4 | 884 | 106 | |
BC141884.1 | - | 4 - 5 | 976 | 106 |