The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003245.3 | NP_003236.3 | 3 - 4 | 517 | 74 |
GenBank mRNA | AK290324.1 | - | 3 - 4 | 481 | 74 |
AK291351.1 | - | 3 - 4 | 481 | 74 | |
AK298637.1 | - | 3 - 4 | 483 | 74 | |
AK308249.1 | - | 3 - 4 | 483 | 74 | |
AK315236.1 | - | 3 - 4 | 481 | 74 | |
BC109075.1 | - | 3 - 4 | 443 | 74 | |
BC109076.1 | - | 3 - 4 | 443 | 74 | |
DC373633.1 | - | 3 - 4 | 517 | 74 |