The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003245.3 | NP_003236.3 | 8 - 9 | 1182 | 75 |
GenBank mRNA | AK290324.1 | - | 8 - 9 | 1146 | 75 |
AK291351.1 | - | 8 - 9 | 1146 | 75 | |
AK298637.1 | - | 8 - 9 | 1148 | 75 | |
AK299447.1 | - | 7 - 8 | 883 | 75 | |
AK315236.1 | - | 8 - 9 | 1146 | 75 | |
BC109075.1 | - | 8 - 9 | 1108 | 75 | |
BC109076.1 | - | 8 - 9 | 1108 | 75 | |
L10386.1 | - | 8 - 9 | 1126 | 75 |