Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBF4
Entrez Gene ID
10926
Gene Name
DBF4 zinc finger
Gene Aliases
ASK, CHIF, DBF4A, ZDBF1
Chromosome Location
Chr.7: 87876229 - 87909541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318060.1 NP_001304989.1 4 - 5 953 138
NM_001318061.1 NP_001304990.1 4 - 5 861 138
NM_001318062.1 NP_001304991.1 4 - 5 953 138
NM_006716.3 NP_006707.1 4 - 5 953 138
GenBank mRNA AB028069.1 - 4 - 5 966 138
AB028070.1 - 4 - 5 966 138
AF160249.1 - 4 - 5 665 138
AF160876.1 - 4 - 5 648 138
AK292569.1 - 4 - 5 695 138
AK302010.1 - 4 - 5 582 138
AK303175.1 - 4 - 5 557 138
BC022811.1 - 4 - 5 550 138
BC036045.1 - 4 - 5 759 138
BC047693.1 - 4 - 5 634 138

View Full Product Details