Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NETO2
Entrez Gene ID
81831
Gene Name
neuropilin and tolloid like 2
Gene Aliases
BTCL2, NEOT2
Chromosome Location
Chr.16: 47081520 - 47144025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721289.1 XP_006721352.1 7 - 8 1293 119
XM_006721291.1 XP_006721354.1 7 - 8 1272 119
GenBank mRNA AK027630.1 - 1 - 2 34 119

View Full Product Details