The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002801.3 | NP_002792.1 | 2 - 3 | 270 | 60 |
GenBank mRNA | AK225658.1 | - | 2 - 3 | 243 | 60 |
AK298012.1 | - | 1 - 2 | 467 | 60 | |
AK312208.1 | - | 2 - 3 | 230 | 60 | |
BC017198.1 | - | 2 - 3 | 221 | 60 | |
BC052369.1 | - | 2 - 3 | 208 | 60 | |
BM845639.1 | - | 2 - 3 | 243 | 60 | |
BP289065.1 | - | 2 - 3 | 270 | 60 | |
BT019723.1 | - | 2 - 3 | 142 | 60 | |
BT019724.1 | - | 2 - 3 | 142 | 60 | |
CR456982.1 | - | 2 - 3 | 142 | 60 | |
Y13640.1 | - | 2 - 3 | 230 | 60 |