Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSF2
Entrez Gene ID
3298
Gene Name
heat shock transcription factor 2
Gene Aliases
HSF 2, HSTF 2
Chromosome Location
Chr.6: 122399551 - 122433119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135564.1 NP_001129036.1 8 - 9 1010 150
NM_004506.3 NP_004497.1 8 - 9 1010 150
GenBank mRNA AK225593.1 - 8 - 9 936 150
AK225594.1 - 8 - 9 936 150
AK294624.1 - 8 - 9 900 150
AK316377.1 - 8 - 9 851 150
BC112323.1 - 8 - 9 913 150
BC121051.1 - 8 - 9 867 150
M65217.1 - 8 - 9 911 150

View Full Product Details