The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198714.1 | NP_942007.1 | 3 - 4 | 1395 | 99 |
GenBank mRNA | AK290634.1 | - | 3 - 4 | 1261 | 99 |
D38298.1 | - | 3 - 4 | 1395 | 99 | |
S69326.1 | - | 3 - 4 | 1360 | 99 | |
X83857.1 | - | 3 - 4 | 1371 | 99 |