Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM29
Entrez Gene ID
23650
Gene Name
tripartite motif containing 29
Gene Aliases
ATDC
Chromosome Location
Chr.11: 120111286 - 120138179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012101.3 NP_036233.2 5 - 6 1558 74
XM_005271490.1 XP_005271547.1 5 - 6 735 74
XM_011542729.1 XP_011541031.1 5 - 6 1582 74
XM_011542732.1 XP_011541034.1 5 - 6 1582 74
XM_011542733.1 XP_011541035.1 5 - 6 735 74
XM_017017453.1 XP_016872942.1 5 - 6 891 74
XM_017017454.1 XP_016872943.1 5 - 6 891 74
GenBank mRNA AF230388.1 - 5 - 6 1558 74
AF230389.1 - 5 - 6 1558 74
AK122720.1 - 6 - 7 1374 74
AK223090.1 - 5 - 6 1516 74
AK299459.1 - 5 - 6 836 74
AK299472.1 - 5 - 6 915 74
AK303969.1 - 5 - 6 796 74
BC010644.1 - 5 - 6 1504 74
BC017352.2 - 5 - 6 1497 74
L24203.1 - 5 - 6 1558 74

View Full Product Details