Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM29
Entrez Gene ID
23650
Gene Name
tripartite motif containing 29
Gene Aliases
ATDC
Chromosome Location
Chr.11: 120111286 - 120138179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012101.3 NP_036233.2 6 - 7 1646 90
XM_005271490.1 XP_005271547.1 6 - 7 823 90
XM_011542729.1 XP_011541031.1 6 - 7 1670 90
XM_011542732.1 XP_011541034.1 6 - 7 1670 90
XM_011542733.1 XP_011541035.1 6 - 7 823 90
XM_017017453.1 XP_016872942.1 6 - 7 979 90
XM_017017454.1 XP_016872943.1 6 - 7 979 90
GenBank mRNA AF230388.1 - 6 - 7 1646 90
AF230389.1 - 6 - 7 1646 90
AK093404.1 - 2 - 3 716 90
AK122720.1 - 7 - 8 1462 90
AK223090.1 - 6 - 7 1604 90
AK299459.1 - 6 - 7 924 90
AK299472.1 - 6 - 7 1003 90
AK303969.1 - 6 - 7 884 90
BC010644.1 - 6 - 7 1592 90
BC017352.2 - 6 - 7 1585 90
L24203.1 - 6 - 7 1646 90

View Full Product Details